Skip to Content
Merck
Silicon 2,3-naphthalocyanine dichloride Dye content 85 %

Silicon 2,3-naphthalocyanine dichloride

Silicon 2,3-naphthalocyanine dichloride Dye content 85 %
Synonym(s):
2,3-Naphthalocyaninato-dichlorosilane, Dichloro-2,3-naphthalocyaninatosilane
Empirical Formula (Hill Notation):
C48H24Cl2N8Si
CAS Number:
Molecular Weight:
811.75

Products

Formula Weight
Physical Form
Chemical Composition
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing