
Sennoside C

Empirical Formula (Hill Notation):
C42H40O19
CAS Number:
Molecular Weight:
848.76
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing