Skip to Content
Merck
(S)-(−)-2-Bromopropionic acid 99% (sum of enantiomers, GC)

(S)-(−)-2-Bromopropionic acid

(S)-(−)-2-Bromopropionic acid 99% (sum of enantiomers, GC)
Linear Formula:
CH3CH(Br)CO2H
CAS Number:
Molecular Weight:
152.97

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing