
Rosiglitazone Related Compound A

Synonym(s):
(5Z)-5-{[4-({2-[Methyl(2-pyridinyl)amino]ethyl}oxy)phenyl]methylidene}-1,3-thiazolidine-2,4-dione
Empirical Formula (Hill Notation):
C18H17N3O3S
CAS Number:
Molecular Weight:
355.41
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing