Skip to Content
Merck
Remifentanil hydrochloride ≥97% (HPLC), powder

Remifentanil hydrochloride

Remifentanil hydrochloride ≥97% (HPLC), powder
Synonym(s):
3-[methoxycarbonyl-4-[(1-oxopropyl)phenylamino]1-piperidine]propanoic acid methyl ester hydrochloride, GI-87084B, Remifentanyl, Ultiva
Empirical Formula (Hill Notation):
C20H28N2O5·HCl
CAS Number:
Molecular Weight:
412.91

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing