Skip to Content
Merck
(R)-(−)-2,2-Dimethyl-1,3-dioxolane-4-methanol 98%

(R)-(−)-2,2-Dimethyl-1,3-dioxolane-4-methanol

(R)-(−)-2,2-Dimethyl-1,3-dioxolane-4-methanol 98%
Synonym(s):
2,3-Isopropylidene-sn-glycerol, L-α,β-Isopropylideneglycerol, L-2,3-O-isopropylidene-sn-glycerol
Empirical Formula (Hill Notation):
C6H12O3
CAS Number:
Molecular Weight:
132.16

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing