
p-Tolylmagnesium bromide solution

Synonym(s):
4-Methylphenylmagnesium bromide
Linear Formula:
CH3C6H4MgBr
CAS Number:
Molecular Weight:
195.34
Products
Boiling Point (°C)
Formula Weight
Markush Class
Markush Group
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing