
Prazosin Related Compound D

Synonym(s):
1-(2-Furoyl)piperazine, 2-Furoic piperazide
Empirical Formula (Hill Notation):
C9H12N2O2
CAS Number:
Molecular Weight:
180.20
Products
Brand
Formula Weight
Melting Point (°C)
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing