
Potassium bitartrate

Synonym(s):
Potassium L-tartrate monobasic, L-(+)-Tartaric acid monopotassium salt, Potassium hydrogen L-tartrate
Empirical Formula (Hill Notation):
C4H5KO6
CAS Number:
Molecular Weight:
188.18
Products
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing