
Phosphatidylethanolamine (Soy)

Synonym(s):
Phosphatidylethanolamines, soya
CAS Number:
EC Number:
Products
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing