
Palmitic-d31 acid

Synonym(s):
Palmitic acid-d31, Hexadecanoic-d31 acid
Linear Formula:
CD3(CD2)14CO2H
CAS Number:
Molecular Weight:
287.62
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Functional Group
Isotope
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing