
MTT Formazan

Synonym(s):
1-(4,5-Dimethylthiazol-2-yl)-3,5-diphenylformazan, Thiazolyl blue formazan
Empirical Formula (Hill Notation):
C18H17N5S
CAS Number:
Molecular Weight:
335.43
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing