
Midazolam

Synonym(s):
8-Chloro-6-(2-fluorophenyl)-1-methyl-4H-imidazo[1,5-a][1,4]benzodiazepine
Empirical Formula (Hill Notation):
C18H13ClFN3
CAS Number:
Molecular Weight:
325.77
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing