Skip to Content
Merck
Methylene Blue hydrate meets analytical specification of BP 73

Methylene Blue hydrate

Methylene Blue hydrate meets analytical specification of BP 73
Synonym(s):
Methylene Blue hydrate
Empirical Formula (Hill Notation):
C16H18ClN3S · xH2O
CAS Number:
Molecular Weight:
319.85 (anhydrous basis)
EC Number:

Products

Product Category
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Technique