Skip to Content
Merck
Mercury(II) oxide red ACS reagent, ≥99.0%

Mercury(II) oxide red

Mercury(II) oxide red ACS reagent, ≥99.0%
Synonym(s):
Mercuric oxide red, Mercury(II) oxide
Linear Formula:
HgO
CAS Number:
Molecular Weight:
216.59

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Core
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing