
Meloxicam impurity B

Synonym(s):
2-Amino-5-methylthiazole
Empirical Formula (Hill Notation):
C4H6N2S
CAS Number:
Molecular Weight:
114.17
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing