
Lauric acid

Synonym(s):
Dodecanoic acid, ABL, Lauric acid, NSC 5026
Linear Formula:
CH3(CH2)10COOH
CAS Number:
Molecular Weight:
200.32
Products
Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-6 of 6
Compare
Product Number
Description
Pricing