
Isobutyryl-L-carnitine

Synonym(s):
(2R)-3-Carboxy-N,N,N-trimethyl-2-(2-methyl-1-oxopropoxy)-1-propanaminium inner salt, L-Carnitine isobutyryl ester, iC4-Carnitine
Empirical Formula (Hill Notation):
C11H21NO4
CAS Number:
Molecular Weight:
231.29
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing