Skip to Content
Merck
Hygromycin B solution from Streptomyces hygroscopicus ≥60% (HPLC), 45-60 mg/mL in H2O, γ-irradiated

Hygromycin B solution from Streptomyces hygroscopicus

Hygromycin B solution from Streptomyces hygroscopicus ≥60% (HPLC), 45-60 mg/mL in H2O, γ-irradiated
Empirical Formula (Hill Notation):
C20H37N3O13
CAS Number:
Molecular Weight:
527.52

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Activity Spectrum
Quality Segment
Mode of Action
Sterilization Method
Showing 1-2 of 2