
Glycerol-13C3

Synonym(s):
1,2,3-Propanetriol-13C3, 13C Labeled glycerol, Glycerin-13C3
Linear Formula:
(HO13CH2)213CHOH
CAS Number:
Molecular Weight:
95.07
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing