
Ginsenoside Rf

Synonym(s):
(3β,6α,12β)-3,12,20-Trihydroxydammar-24-en-6-yl 2-O-β-D-glucopyranosyl-β-D-glucopyranoside, Panaxoside Rf
Empirical Formula (Hill Notation):
C42H72O14
CAS Number:
Molecular Weight:
801.01
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing