Skip to Content
Merck
Giemsa stain technical grade, used as a blood stain

Giemsa stain

Giemsa stain technical grade, used as a blood stain
Synonym(s):
Azure mixture sicc. Giemsa stain
Empirical Formula (Hill Notation):
C14H14ClN3S
CAS Number:
Molecular Weight:
291.80

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Technique
Showing 1-2 of 2