
FEBANTEL

Synonym(s):
Febantel, N-{2-[2,3-Bis-(methoxycarbonyl)-guanido]-5-(phenylthio)-phenyl}-2-methoxyacetamide
Empirical Formula (Hill Notation):
C20H22N4O6S
CAS Number:
Molecular Weight:
446.48
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3