
FD&C Yellow No. 6

Synonym(s):
Sunset Yellow FCF, Food Yellow 3, Orange Yellow S
Empirical Formula (Hill Notation):
C16H10N2Na2O7S2
CAS Number:
Molecular Weight:
452.37
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing