Skip to Content
Merck
vibrant-m

Fast green FCF (C.I. 42053)

vibrant-m
Empirical Formula (Hill Notation):
C37H34N2Na2O10S3
CAS Number:
Molecular Weight:
808.85

Products

Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing