
EZview™ Red ANTI-FLAG® M2 Affinity Gel

Synonym(s):
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, Anti-ddddk, Anti-dykddddk
Products
Brand
Clonality
Isotype
Analyte Suitability
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing