Ethyl oleate
Synonym(s):
Oleic acid ethyl ester
Linear Formula:
CH3(CH2)7CH=CH(CH2)7COOC2H5
CAS Number:
Molecular Weight:
310.51
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Showing 1-5 of 5