Skip to Content
Merck
Ethyl maltol analytical standard

Ethyl maltol

Ethyl maltol analytical standard
Synonym(s):
2-Ethyl-3-hydroxy-4H-pyran-4-one, Ethyl maltol
Empirical Formula (Hill Notation):
C7H8O3
CAS Number:
Molecular Weight:
140.14
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing