
Ethylenediaminetetraacetic acid solution

Synonym(s):
EDTA solution
Empirical Formula (Hill Notation):
C10H16N2O8
CAS Number:
Molecular Weight:
292.24
Products
Brand
Formula Weight
Physical Form
Product Line
Sterilization Method
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing