
Ethanol-2-13C

Synonym(s):
13C Labeled ethanol, Ethyl alcohol-2-13C
Linear Formula:
13CH3CH2OH
CAS Number:
Molecular Weight:
47.06
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing