
(±)-Methoxyverapamil hydrochloride

Synonym(s):
(±)-α-[3-[(2-(3,4-Dimethoxyphenyl)ethyl)methylamino]propyl]-3,4,5-trimethoxy-α-(1-methylethyl)benzeneacetonitrile hydrochloride, D 600, Gallopamil
Empirical Formula (Hill Notation):
C28H40N2O5 · HCl
CAS Number:
Molecular Weight:
521.09
Products
Product Category
Brand
Formula Weight
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing