Skip to Content
Merck
Di-tert-butylsilyl bis(trifluoromethanesulfonate) 97%

Di-tert-butylsilyl bis(trifluoromethanesulfonate)

Di-tert-butylsilyl bis(trifluoromethanesulfonate) 97%
Synonym(s):
DTBS ditriflate, Di-tert-butylsilyl ditriflate, Trifluoromethanesulfonic acid di-tert-butylsilylene ester
Linear Formula:
(CF3SO3)2Si[C(CH3)3]2
CAS Number:
Molecular Weight:
440.45

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing