
Dextroamphetamine sulfate

Synonym(s):
D-Amphetamine hemisulfate salt, (+)-α-Methylphenethylamine hemisulfate salt, Dextroamphetamine hemisulfate salt
Empirical Formula (Hill Notation):
C9H13N · 0.5 H2SO4
CAS Number:
Molecular Weight:
184.25
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing