
Deltamedrane

Synonym(s):
(6α,11β)-11,17-Dihydroxy-6-methylpregna-1,4-diene-3,20-dione, 11β,17α-Dihydroxy-6α-methylpregna-1,4-diene-3,20-dione
Empirical Formula (Hill Notation):
C22H30O4
CAS Number:
Molecular Weight:
358.47
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing