
Dehydroacetic acid

Synonym(s):
2-Acetyl-5-hydroxy-3-oxo-4-hexenoic acid δ-lactone, Dehydroacetic acid
Empirical Formula (Hill Notation):
C8H8O4
CAS Number:
Molecular Weight:
168.15
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique