
Cromolyn Related Compound B

Synonym(s):
Diethyl 5,5′-[(2-hydroxypropane-1,3-diyl)bis(oxy)]bis(4-oxo-4H-chromene-2-carboxylate), Diethyl cromoglycate
Empirical Formula (Hill Notation):
C27H24O11
CAS Number:
Molecular Weight:
524.47
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2