
Creatinine

Synonym(s):
2-Amino-1-methyl-2-imidazolin-4-one, 2-Imino-1-methylimidazolidin-4-one, 2-Imino-N-methylhydantoin, Creatinine
Empirical Formula (Hill Notation):
C4H7N3O
CAS Number:
Molecular Weight:
113.12
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4