
Chlorhexidine digluconate

Synonym(s):
Chlorhexidine digluconate solution, 1,6-Bis(N5-[p-chlorophenyl]-N1-biguanido)hexane; 1,1′-Hexamethylenebis(5-[p-chlorophenyl]biguanide)
CAS Number:
Products
Brand
Agency
Format
Quality Segment
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing