Skip to Content
Merck
Cerium(III) nitrate hexahydrate 99% trace metals basis

Cerium(III) nitrate hexahydrate

Cerium(III) nitrate hexahydrate 99% trace metals basis
Synonym(s):
Cerium trinitrate, Cerous nitrate hexahydrate, Nitric acid cerium salt, Cerium(III) nitrate hexahydrate
Linear Formula:
Ce(NO3)3 · 6 H2O
CAS Number:
Molecular Weight:
434.22

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Reagent Type