
Cefixime

Synonym(s):
Cefixime trihydrate
Empirical Formula (Hill Notation):
C16H15N5O7S2 · 3H2O
CAS Number:
Molecular Weight:
507.50
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing