
Cardiolipin solution from bovine heart

Synonym(s):
Cardiolipin solution from bovine heart
EC Number:
Products
Product Category
Brand
Biological Source
Color
Physical Form
Purity
Research Area
Tissue
Quality Segment
Functional Group
Application
Package Size