
Calcium saccharate

Synonym(s):
Calcium D-saccharate tetrahydrate, D-Glucaric acid calcium salt, D-Saccharic acid calcium salt, Calcium D-glucarate
Empirical Formula (Hill Notation):
C6H8CaO8 · 4H2O
CAS Number:
Molecular Weight:
320.26
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing