Skip to Content
Merck
Bis(2-ethylhexyl) adipate Selectophore™, ≥99.0%

Bis(2-ethylhexyl) adipate

Bis(2-ethylhexyl) adipate Selectophore™, ≥99.0%
Synonym(s):
Adipic acid di(2-ethylhexyl) ester, DOA, Bis(2-ethylhexyl) adipate
Linear Formula:
[-CH2CH2CO2CH2CH(C2H5)(CH2)3CH3]2
CAS Number:
Molecular Weight:
370.57
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique