Skip to Content
Merck
Betulinic acid analytical standard

Betulinic acid

Betulinic acid analytical standard
Synonym(s):
3β-Hydroxy-20(29)-lupaene-28-oic acid, Lupatic acid, Mairin
Empirical Formula (Hill Notation):
C30H48O3
CAS Number:
Molecular Weight:
456.70
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4