
Anti-Pig IgG (whole molecule)−FITC antibody produced in rabbit

Synonym(s):
Anti-Porcine IgG (whole molecule)−FITC antibody produced in rabbit, Anti-Swine IgG (whole molecule)−FITC antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Technique