Anti-MAP2 Antibody
Synonym(s):
Anti-Microtubule Associated Protein 2, Anti-Microtubule associated protein 2, MAP2A, MAP2B, MAP2C, Anti-MAP 2, Anti-MAP-2, Anti-Microtubule-associated protein 2
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-16 of 16
Compare
Product Number
Description
Pricing