
Anti-Human IgG2−FITC antibody, Mouse monoclonal

Synonym(s):
Monoclonal Anti-Human IgG2
Products
Product Category
Brand
Biological Source
Clonality
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Post Translational Modification
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing