
Amisulbrom

Synonym(s):
3-(3-Bromo-6-fluoro-2-methylindol-1-ylsulfonyl)-N,N-dimethyl-1H-1,2,4-triazole-1-sulfonamide
Empirical Formula (Hill Notation):
C13H13BrFN5O4S2
CAS Number:
Molecular Weight:
466.31
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing