
Aminolevulinic acid Related Compound A

Synonym(s):
Pyrazine-2,5-dipropionic acid, 3,3′-(Pyrazine-2,5-diyl)dipropionic acid
Empirical Formula (Hill Notation):
C10H12N2O4
CAS Number:
Molecular Weight:
224.21
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2