
Amikacin for system suitability

Synonym(s):
Amikacin disulfate salt
Empirical Formula (Hill Notation):
C22H43N5O13 · 2H2SO4
CAS Number:
Molecular Weight:
781.76
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing